Would you like to log in again?
EOTAXIN/CCL11 HUMAN
Supplier: Genscript
EOTAXIN/CCL11 HUMAN^ CCL11 is a potent eosinophil chemoattractant that was originally purified from Bronchoalveolar lavage fluid of guinea pigs sensitized By aerosol challenge with ovalBumin. Human CCL11 cDNA encodes a 97 a.a residue precursor protein from which the aminoterminal 23 a.a residues
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EOTAXIN/CCL11 HUMAN 89306-042 Z02835-20 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty