Would you like to log in again?
CXCL16 MOUSE
Supplier: Genscript
CXCL16 MOUSE^ Mouse CXC chemokine 16 is a nonELR motifcontaining CXC chemokine with a transmemBrane domain. CX3CL1/Fractalkine and CXCL16 are the only two transmemBrane chemokines within the superfamily. Mouse CXCL16 cDNA encodes a 246 amino acid (aa) precursor protein with a putative 26 aa
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CXCL16 MOUSE 89306-112 Z02853-1 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty