Would you like to log in again?
INTERFERON-A1B IFN-A1B HUMAN
Supplier: Genscript
INTERFERON-A1B IFN-A1B HUMAN^ The individual proteins have molecular masses Between 19-26 kDa and consist of proteins with lengths of 156-166 and 172 amino acids. IFN-alpha suBtypes possess a common conserved sequence region Between amino acid positions115-151
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INTERFERON-A1B IFN-A1B HUMAN 89306-160 Z02866-10 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty