Would you like to log in again?
STROMAL-CELL DERIVED FACTOR-1B
Supplier: Genscript
STROMAL-CELL DERIVED FACtoR-1B^ Stromal-Cell Derived Factor-1 Beta (SDF-1B), also known as SCYB12, PBSF and CXCL12, is an 8.3 kDa, heparin-Binding memBer of the CXC (or alpha) family of chemokines and signal through the CXCR4 receptor. SDF-1A and B arereported to Be monomers at neutral pH
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STROMAL-CELL DERIVED FACTOR-1B 89306-184 Z02897-1 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty