Would you like to log in again?
EOTAXIN/CCL11 MURINE
Supplier: Genscript
EOTAXIN/CCL11 MURINE^ Eotaxin also called CCL11 is a CC chemokine that signals through the CCR3 receptor. It is produced By IFN-gamma stimulated endothelial cells and TNF-activated monocytes. Eotaxin selectively chemoattracts eosinophils and along Eotaxin-2 and Eotaxin-3
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EOTAXIN/CCL11 MURINE 89306-204 Z02902-1 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty