Would you like to log in again?
INTERFERON-G IFN-G MOUSE
Supplier: Genscript
INTERFERON-G IFN-G MOUSE^ Sharing 41% sequence identity with human Interferon gamma (hIFN-G), mouse IFN gamma (mIFN-G) is a macrophage-activating factor.The active form of IFN-G is an antiparallel dimer that sets off IFN-G/JAK/STAT pathway.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INTERFERON-G IFN-G MOUSE 89306-260 Z02916-20 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty