Would you like to log in again?
Tris-Buffered Saline with Tween Solution (10X), Genetex
Supplier: GeneTex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GTX30976
89306-978EA 114.12 USD InStock
89306-978
Tris-Buffered Saline with Tween Solution (10X), Genetex
Buffers Electrophoresis Buffers Washing Buffers
Dilute this 10X buffer 10 times, mix well, and store 1 X buffer solution at room temperature or 2-8oC
Liquid form. Store at room temperature. If stored at 2°C-8°C, some sodium chloride will be precipitated, if this occurs, bring the buffer to room temperature and mix well till all sodium chloride is dissolved.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty