Would you like to log in again?
FluoroGel Mounting Medium, Genetex
Supplier: GeneTex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Fluoro-Gel
GTX28214
89336-416EA 320.04 USD InStock
89336-416
FluoroGel Mounting Medium, Genetex
Stains Microscopy and staining reagents
FluoroGel is an aqueous-based mounting medium designed for the preservation of fluorescence-stained tissue sections.
  • For ICC/IF, IHC, IHC-Fr, IHC-P
  • Storage Temperature 4 °C
FluoroGel is a non-glycerol based mounting medium. It is specially formulated without glycerol to avoid the deleterious effects of this compound in some of the phycobili-protein based detection systems. This makes FluoroGel the mounting medium of choice when phycobili proteins are used as tracers. This product is not suitable for binary substrates generally used for immunohistochemistry. A product like Clear Mount (aqueous mounting media) or Limonene (organic mounting media) should be used.
Can also be used with conventional fluorescent tracers such as fluorescein (FITC), Rhodamine (TMRITC) and Texas Red.
Caution: For In vitro laboratory use only. Not for any clinical, therapeutic, or diagnostic use in humans or animals. Not for animal or human consumption.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty