Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CPN10 Mouse Monoclonal Antibody [clone: M1.2]

Supplier: GeneTex
GTX13527
89359-726EA 609.52 USD InStock
89359-726
Anti-CPN10 Mouse Monoclonal Antibody [clone: M1.2]
Antibodies
Homologs of E. coli GroES make up a family of small heat shock proteins with an approximate molecular mass of 10 kDa (Hsp10s). Cpn10 acts as a cochaperone and interacts with members of the Hsp60 family (GroEL in E. coli) to promote proper folding of polypeptides. In chlamydia, Cpn10 is thought to interact with cHsp60, a reported immunopathological antigen, identifying cpn10 as a potential contributor to an immunological response.

EIA: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a concentration of 0.25 ?g/ml. Detects a band of approximately 15 kDa (predicted molecular weight: 9.9 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CPN10
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty