Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADAMTS5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX16314
89360-210EA 604.89 USD InStock
89360-210
Anti-ADAMTS5 Rabbit Polyclonal Antibody
Antibodies
ADAMTS5, also known as Implantin or Aggrecanase 2, is a member of the larger family of ADAMs (A Disintegrin And Metalloproteinase) metalloproteinases containing thrombospondin (TS) repeats. ADAMTS5 (A Disintegrin And Metalloproteinase with Thrombospondin-5 motif) was first described as ADAMTS5, a protein elevated in mice during the peri-implantion period. At the same time, another group identified Aggrecanase 11, a protein elevated in arthritic synovium. The name was later changed to ADAMTS5. ADAMTS5 is expressed in human and mouse. It has been found in heart, lung, cervix, uterus, ovary, brain, cartilage, and numerous other tissues, as well as chondroblastoma cell lines. Initial observations indicated a role for ADAMTS5 in aggrecan cleavage and cartilage destruction, especially in arthritis and potentially a role in embryo implantion. ADAMTS5 contains the canonical HExxHxxxxxH zinc metalloproteinase motif, and has been shown to efficiently cleave Aggrecan. In addition to the metalloprotease domain, ADAMTS5 has a propeptide domain, a prohormone convertase (PC, furin) cleavage site, a cysteine-rich domain, a spacer domain, and two thrombospondin-1 like domains. ADAMTS5 is inhibited by the endogenous MMP inhibitors (TIMP1, 2, 3, and 4) but most efficiently by TIMP3. Unlike many of the ADAMs proteases, ADAMTS5 does not have a transmembrane domain, and is a secreted protein. Full length ADAMTS5 is a 930 amino acid protein with a predicted molecular mass is 101.7 kDa, but glycosylation and the abundance of cysteine residues gives ADAMTS5 a greater apparent molecular weight on reduced SDS PAGE gels. When ADAMTS5 is secreted, it is cleaved at the furin cleavage site (predicted molecular mass 73.2 kDa) and then further cleaved to generate a range of smaller forms.

Recommended Starting Dilutions:IHC: 4 ug/mlWB: 1:1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ADAMTS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty