Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL6 Goat Polyclonal Antibody

Supplier: GeneTex
GTX17529
89360-314EA 602.26 USD InStock
89360-314
Anti-IL6 Goat Polyclonal Antibody
Antibodies
Interleukin 6 (IL6) is a multifunctional 24 kD protein originally discovered in the medium of RNA stimulated fibroblastoid cells. It is upregulated by IL1, TNF, PDGF, IFN beta, TNF alpha, NGF, IL17 and downregulated by glucocorticoids IL4, TGF beta. IL6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be as important as IL1 and TNF alpha in regulating the acute phase response. IL6 has also been implicated in regulating adipose mass. IL6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. It acts upon a variety of cells including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. In addition, IL6 appears to interact with IL2 in the proliferation of T lymphocytes. IL6 potentiates the proliferative effect of IL3 on multipotential hematopoietic progenitors.

ELISA: Use at a concentration of 0.15 - 0.3 ?g/ml. This concentration allows the detection of at least 0.2 ng/well of recombinant hIL6. WB: Use at a concentration of 0.1 - 0.2 ?g/ml. The detection limit for recombinant hIL6 is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions. Predicted molecular weight: 26 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty