Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EPO Mouse Monoclonal Antibody [clone: 85]

Supplier: GeneTex
GTX19485
89360-618EA 611.88 USD InStock
89360-618
Anti-EPO Mouse Monoclonal Antibody [clone: 85]
Antibodies
Human erythropoietin is an acidic glycoprotein hormone with a molecular mass of 34 kD. As the prime regulator of red cell production, its major functions are to promote erythroid differentiation and to initiate hemoglobin synthesis.

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: EPO
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty