Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD24 Mouse Monoclonal Antibody [clone: ALB9]

Supplier: GeneTex
GTX19704
89360-670EA 617.6 USD InStock
89360-670
Anti-CD24 Mouse Monoclonal Antibody [clone: ALB9]
Antibodies
CD24 is a glycoprotein with a polypeptide core of 30 amino acids and high carbohydrate content. The apparent molecular mass of the two forms of CD24 purified from a mouse lymphoma cell line appear to be in the range of 40-60 and 23-30 kDa. The CD24 antigen is present on lymphocytes, granulocytes, erythrocytes.

FACS: 2ug/5x100000cells/test. Immunohistochemistry (frozen sections or cell smears): 1/25 - 1/50. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: ALB9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty