Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SSEA5 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX70019
89364-654EA 582.76 USD InStock
89364-654
Anti-SSEA5 Mouse Monoclonal Antibody
Antibodies
Stage-specific embryonic antigen (SSEA)-5 is an H type-1 glycan that is highly and specifically expressed on human embryonic stem cells (hESCs), the human blastocyst inner cell mass and undifferentiated human pluripotent stem cells (hPSCs). Expression of SSEA-5 in hESCs and hPSCs rapidly decreases upon differentiation and is considered an indicator of undifferentiated PSCs. This SSEA-5 antibody is capable of identifying undifferentiated pluripotent stem cells in heterogeneous populations, and when used in conjunction with other pluripotency surface markers (CD9 and CD90 or CD50 and CD200), it can be used to immunodeplete residual teratoma-causing cells from incompletely differentiated embryonic stem cell cultures.

Type: Primary
Antigen: SSEA5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty