Would you like to log in again?
VWR® Traceable® Platinum High-Accuracy Freezer Thermometer
Supplier: VWR
90009-83 90013-57
89369-152EA 282.28 USD InStock
89369-152 89369-150
VWR® Traceable® Platinum High-Accuracy Freezer Thermometer
Thermometers Digital Thermometers
Thermometer displays minimum/maximum temperatures for last 24 hours for each hour and previous 7 days for each days; or last 30 days for each day.
  • Ideal for freezers
  • Displays exact time and date for all readings
  • Highly accurate across entire temperature range
  • Features a waterproof cable
  • Stainless steel probe
Time/date stamp key shows the exact time and date for all minimum and maximum readings
Supplied platinum waterproof stainless steel probe provides accruate readings with tip penetration of 0.3", waterproof cable, and adjustable probe holder or supplied with platinum waterproof stainless steel Bullet™ probe and waterproof cable. Handle probe model features stainless steel probe while Bullet(TM) model features PTFE cable to withstand temperatures of −100 to +200 °C. Features a 10-foot waterproof cable.
A 4-wire platinum sensor insures linear response and high accuracy across the entire temperature range. Thermometer is ideal for freezers.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Ordering information: Supplied with platinum probe/cable, wall mount, bench stand, batteries, Traceable® certificate.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to VWR's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty