Would you like to log in again?
VWR® Traceable® Extra-Extra Large Digit Timer
Supplier: Avantor
VWR®
93678-96
89369-164EA 44.3 USD InStock
89369-164
VWR® Traceable® Extra-Extra Large Digit Timer
Timers Digital Timers
Extra large 1¹/₂" digit size allows for easy viewing from up to 60 feet away.
  • Easy to use
  • Automatic timing memory
  • Large, easy-to-read display
Reliable, 3-key operation makes countdown settings in 1 second increments easy to program. Timer countdowns and alarms on single channel for ease-of-use. Counts up during alarm function indicating elapsed time since alarming. Times up to 99 minutes, 59 seconds with one second resolution. Automatic Bounceback(TM) Memory for repetitive timing. Accuracy is 0.01%. Counts up in stopwatch mode.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Ordering information: Supplied with wall mount clip, bench stand, magnetic back, battery and Traceable® Certificate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty