Would you like to log in again?
Shandon™ Bluing Reagent, Epredia
Supplier: Epredia
Shandon
6769001 6769002 6769001 6769002
89369-346EA 111.69 USD InStock
CA89369-346 CA89369-344 89369-346 89369-344
Shandon™ Bluing Reagent, Epredia
Stains Microscopy and staining reagents
Employ a gentle, pH-controlled solution for bluing hematoxylin with the Epredia™ Shandon™ Bluing Reagent.
  • Ready-to-use
  • Thoroughly pH-controlled for completely effective bluing
It replaces the strong alkaline rinses or long tap water washes which may cause loss of cells or tissue sections.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty