Would you like to log in again?
MEGA FlexTubes, IBI Scientific
Supplier: IBI Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IB48441 IB48302 IB48340 IB48321 IB48431 IB48370 IB48342 IB48310 IB48301 IB48331 IB48007 IB48410 IB48320 IB48422 IB48391 IB48361 IB48360 IB48390 IB48300 IB48371 IB48311 IB48330 IB48450 IB48420 IB48440 IB48451 IB48400 IB48381 IB48382 IB48350 IB48401 IB48430 IB48380 IB48351 IB48341 IB48411 IB48421
89369-612PK 48.5 USD InStock
89369-612 89369-656 89369-634 89369-610 89369-456 89369-632 89369-654 89369-598 89369-652 89369-630 89369-596 89369-650 89369-594 89369-608 89369-606 89369-628 89369-626 89369-604 89369-648 89369-624 89369-602 89369-646 89369-600 89369-644 89369-622 89369-664 89369-642 89369-620 89369-662 89369-640 89369-660 89369-618 89369-616 89369-638 89369-636 89369-658 89369-614
MEGA FlexTubes, IBI Scientific
Tubes Dialysis Tubes
IBI MEGA FlexTubes are typically used for dialysis or buffer exchange with volumes of 3 to 20 ml.
The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.
IBI FlexTubes allow rapid, secure, and simple loading and recovery of samples. IBI FlexTubes can also be used for applications such as the preparation of protein samples for MALDI-MS, sample concentration, tissue culture extraction purification, removal of salts, surfactants, solvents, and detergents, as well as peptide dialysis and virus-particle purification. The performance of IBI MEGA FlexTubes are regularly monitored and tested by simple dialysis and assays commonly used for proteins and/or nucleic acids.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty