Would you like to log in again?
Eyesaver® Eyewash Faucets, Speakman®
Supplier: Speakman
Eyesaver®
SEF-1850-TW SEF-1850-ST SEF-1850 SEF18504WHTW
89369-692EA 1763.98 USD InStock
89369-692 89369-680 89369-678 89369-686
Eyesaver® Eyewash Faucets, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Dedicated water ways allow the faucet and eyewash to work independently of each other.
  • 'LIFT' activator for eyewash
  • Aerated plastic spray outlets with flip-top dust caps
  • Lead free brass construction
Model SEF-1850 is a single post laboratory faucet with an integrated ANSI compliant aerated eyewash.
Model SEF-1850-TW is a single post laboratory faucet with an integrated ANSI compliant aerated eyewash and tepid water system.
Model SEF-1850-4WH-TW is a single post laboratory faucet with an integrated ANSI compliant aerated eyewash and tepid water system. The faucet is equipped with 4" wrist blade handles.
Model SEF-1850-ST is a single post laboratory faucet with vacuum breaker and serrated tip. Includes an integrated ANSI compliant aerated eyewash.
Certifications: Meets ANSI Z358.1, ASME A112.18.1/CSA B125.1, NSF 61, CA AB 1953 and VT Act 193.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty