Would you like to log in again?
Super Adjustable 2™ Super Erecta Shelf® Wire Shelving, Metroseal 3™ Finish, Metro
Supplier: Metro International
Super Adjustable™
A2160NK3 A1860NK3 A2142NK3 A1854NK3 A2154NK3 A2124NK3 A1872NK3 A1448NK3 A1448NK3 A2436NK3 A2172NK3 A1836NK3 A2430NK3 A2448NK3 A2172NK3 A2424NK3 A1854NK3 A2454NK3 A2142NK3 A1836NK3 A2424NK3 A2442NK3 A2148NK3 A2448NK3 A2124NK3 A2154NK3 A1842NK3 A1430NK3 A2130NK3 A1472NK3 A1848NK3 A2442NK3 A1460NK3 A2136NK3 A2430NK3 A2454NK3 A2130NK3 A1860NK3 A2460NK3 A1830NK3 A2148NK3 A1842NK3 A1472NK3 A1424NK3 A1460NK3 A1848NK3 A2472NK3 A1436NK3 A2460NK3 A1442NK3 A1830NK3 A2160NK3 A2136NK3 A1436NK3 A2472NK3 A1824NK3 A1424NK3 A1442NK3 A1824NK3 A2436NK3 A1430NK3 A1872NK3
83009-430EA 149.92 USD InStock
CA22233-773 83009-430 CA83009-440 83009-432 CA83009-442 83009-434 CA83009-444 83009-436 CA89369-840 CA83009-446 89369-798 83009-438 CA89369-762 CA83009-448 89369-792 89369-828 CA89369-834 CA89369-756 CA89369-798 89369-822 CA83009-430 CA83009-432 CA83009-434 CA89369-774 CA83009-436 89369-840 89369-762 CA83009-438 CA89369-804 CA89369-768 89369-834 89369-756 83009-450 CA83009-460 83009-452 83009-454 83009-456 CA89369-786 83009-458 89369-810 89369-774 CA89369-780 CA89369-816 89369-804 CA89369-810 89369-768 83009-460 89369-780 83009-440 CA83009-450 83009-442 CA83009-452 83009-444 83009-446 CA83009-456 83009-448 CA83009-458 89369-786 CA89369-792 89369-816 CA89369-822 CA89369-828
Super Adjustable 2™ Super Erecta Shelf® Wire Shelving, Metroseal 3™ Finish, Metro
Furniture Shelves
Metroseal 3 Corrosion-resistant shelves are electroplated carbon steel, with a special epoxy coating.
  • Metroseal 3™ shelves are an economical alternative to stainless steel
  • Metroseal 3™ shelves includes antimicrobial product protection, which inhibits the growth of bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation
  • Use within coolers to resist corrosion from moisture
  • Shelves are not cart-washable
Open-wire design minimizes dust accumulation, allows a free circulation of air, and provides greater visibility of stored items. Shelves up to 121.9 cm (48") in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") and all super wide shelves can support up to 272 kg (600 lbs.) per level.
Certifications: Shelves meet MIL-S-40144E specifications.
Ordering information: Each shelf and post is sold separately. To order posts or accessories, contact your Avantor representative for more information. Each shelf includes four pairs of plastic split sleeves for securing shelf to posts. Manufacturer's 12-year limited warranty against rust formation.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty