Would you like to log in again?
Super Adjustable 2™ Super Erecta Shelf® Wire Shelving, Stainless Steel, Metro
Supplier: Metro International
Super Adjustable™
A2130NS A2454NS A2136NS A1842NS A1430NS A2160NS A2424NS A1436NS A1860NS A1872NS A2454NS A1830NS A1460NS A2124NS A2148NS A2136NS A1430NS A1854NS A2472NS A1842NS A2172NS A2130NS A1848NS A2160NS A1854NS A1442NS A1872NS A1460NS A2424NS A2172NS A2430NS A1836NS A1830NS A2448NS A2436NS A2448NS A2436NS A2154NS A2472NS A2148NS A1860NS A2430NS A1448NS A2442NS A1824NS A2460NS A1848NS A2442NS A1824NS A2154NS A2142NS A1472NS A1448NS A1424NS A1836NS A2124NS A1424NS A1436NS A2142NS A2460NS A1442NS A1472NS
89369-830EA 756.05 USD InStock
CA89369-764 89369-830 89369-794 CA19766-970 19766-962 CA89369-836 19766-960 CA89369-758 89369-824 19766-968 19766-966 19766-964 CA19766-958 89369-842 89369-764 CA89369-770 19766-950 CA19766-960 89369-836 CA89369-800 89369-758 CA89369-842 19766-958 CA19766-968 19766-956 CA19766-966 19766-954 CA19766-964 19766-952 CA19766-962 CA89369-806 CA19766-948 89369-776 CA89369-782 89369-770 89369-806 19766-940 CA89369-812 89369-800 CA89369-776 CA19766-956 19766-948 CA19766-954 19766-946 CA19766-952 19766-944 CA89369-818 19766-942 CA19766-950 89369-788 CA89369-830 CA89369-794 89369-782 89369-818 19766-970 CA89369-824 89369-812 CA89369-788 CA19766-946 CA19766-944 CA19766-942 CA19766-940
Super Adjustable 2™ Super Erecta Shelf® Wire Shelving, Stainless Steel, Metro
Furniture Shelves
Type 304 stainless steel shelves provide maximum protection in corrosive environments.
  • Adjustable shelves with quick and easy shelf adjustment
  • Design own wire shelf system with appropriate accessories
Notes: The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap. The 245.4 cm high stationary posts are not for use with shelves less than 61 cm wide.
Adjustable shelves feature a unique corner release system that allows for quick and easy shelf adjustment without the use of tools. Shelves are designed for use with SiteSelect® stationary or mobile posts, which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the posts. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Certifications: Units meet MIL-S-40144E specifications.
Ordering information: Each shelf and post are sold separately. All shelves include four pairs of plastic split sleeves for securing shelf to posts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty