Durable chrome-plated shelves are ideal for dry environments with minimal exposure to corrosives.
- Design wire shelf system with appropriate accessories
An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable leveling bolts for use on uneven surfaces. Mobile posts require the purchase of stem casters. Stem casters (see 19766-236 series) and shelf dividers and ledges (see 22233-212 series) are available separately.
Notes: The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap. The 245.4 cm high stationary posts are not for use with shelves less than 61 cm wide.
Adjustable shelves feature a unique corner release system that allows for quick and easy shelf adjustment without the use of tools. Shelves are designed for use with SiteSelect® stationary or mobile posts, which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the posts.
Certifications: Units meet MIL-S-40144E specifications.
Ordering information: Each shelf and post are sold separately. All shelves include four pairs of plastic split sleeves for securing shelf to posts.