Would you like to log in again?
Flexline™ Lab Carts, Metro
Supplier: Metro International
FLTMENT2 FLBED FLCAST FLTMENT2 FLCAST FLISO3 FLISO1 FLISO1 FLBED FLISO3
89369-890EA 5923.73 USD InStock
89369-890 CA89369-888 CA89369-886 89369-888 CA89369-884 89369-886 CA89369-894 89369-884 89369-894 CA89369-890
Flexline™ Lab Carts, Metro
Carts
Flexline enclosed polymer cart system provide unique flexibility and contemporary styling that withstands the rigors of a lab environment.
  • Corrosion-proof, smooth, stain-resistant polymer surfaces
  • Rounded corners make cleaning quick and thorough
  • Optional pull out side shelf provides additional work space
  • Optional laptop arm and keyboard shelf accommodate technology needs
  • Optional backboard can be mounted to the front or back of the cart
  • Optional passive security seals for the entire cart or individual drawers
Microban® antimicrobial product protection is built in every cart, inhibiting the growth of mold, mildew and bacteria
Carts are available in various configurations. Available features also include a pull out interior shelf that accommodates equipment or is used as a seated writing surface, and recessed side storage that holds tilt out bins, wastebaskets, lockable sharps containers, glove boxes, and suction pump shelves.
Full extension drawers have self-closing ball bearing slides to provide easy access to medications and supplies (Drawer depths: 3", 6", 9", 12"). 5" (127mm) polymer caster, 2 directional and 2 total lock casters provide control and stability.
Ordering information: Additional accessories are available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty