Would you like to log in again?
Bow-Tie™ Dunnage Racks, Metro
Supplier: Metro International
HP2248PD HP2236PDMB HP2230PD HP2230PD HP2248PD HP2236PD HP2248PDMB HP2260PDMB HP2230PDMB HP2260PD HP2236PDMB HP2230PDMB HP2260PDMB HP2248PDMB HP2236PD HP2260PD
89369-898EA 526.61 USD InStock
CA89369-896 89369-898 89369-896 CA89369-904 89369-906 89369-904 CA89369-902 89369-902 CA89369-900 CA89369-910 CA89369-898 89369-900 89369-910 CA89369-908 89369-908 CA89369-906
Bow-Tie™ Dunnage Racks, Metro
Furniture Shelves
Heavy-duty construction with rust and corrosion-proof polymer material.
  • Available with built-in Microban® antimicrobial product protection to protect from bacteria, mold, mildew and fungi that cause odors, stains and product degradation
  • Standard models, without Microban are also available
  • Racks join together easily without tools in end-to-end and back-to-back configurations with the exclusive Bow-Tie™ feature
  • Helps protect floors: legs have a larger radius than traditional dunnage racks and distribute the storage load across a wider area
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty