Would you like to log in again?
MetroMax i™ High Performance Liquid Chromatography (HPLC) Carts, Metro™
Supplier: Metro International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MetroMax i™
HPLC2X3 HPLC3X3 HPLC1X3 HPLC3X3 HPLC4X3
82000-920EA 3523.88 USD InStock
82000-920 CA89369-912 89369-912 82000-918 97005-894
MetroMax i™ High Performance Liquid Chromatography (HPLC) Carts, Metro™
Carts
HPLC carts contribute to greater benchtop productivity.
  • Mobilize and stack HPLC equipment on carts to maximize valuable lab space
  • Avoid crowding valuable bench top space with HPLCor other analytical equipment
Five-shelf units are mounted on 188cm (74") posts with casters and are used for stacking and mobilizing HPLC or other analytical equipment
Slide-out keyboard shelf is epoxy-coated for a high level of corrosion resistance. Flat monitor mounting kit features a unique bracket and crossbar design for mounting flat computer monitors between two MetroMax i posts, either in back or front of unit. Six-outlet power strip with surge protection mounts flush with the shelf frame and includes a 1.8m (6') cord. Power strip is UL listed.
They feature strong polymer construction with superior corrosion resistance. Shelves are adjustable in 2.5cm (1") increments along the height of the post. Each five-shelf cart has four solid shelves and one open-grid shelf. Each shelf includes a mat made from mineral-reinforced polypropylene with built-in antimicrobial protection from bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation. Carts can be used in cold conditions as low as –29°C (–20°F).
Ordering information: Manufacturer’s lifetime guarantee against corrosion and rust on shelves and posts. Each five-shelf cart includes four 12.7cm (5") polyurethane casters (two swivel, two brake), four solid shelf mats, and one grid shelf mat. Four-shelf carts are not supplied with casters; see 19766-254 series. Carts are also available in 91.4cm (36") length; contact your Avantor sales representative for more information.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty