Would you like to log in again?
MetroMax i™ Shelves and Posts, Metro™
Supplier: Metro International
MetroMax i® MetroMax i™
MX2472G MX1824F MX1836F MX74P MX1872F MX63UP MX13UP MX1830G MX1830F MX1860G MX1854G MX33UP MX2430G MX1836F MX2442G MX1872G MX2454G MX2454F MX2448G MX1848G MX1824F MX2448F MX1842G MX2424G MX86P MX2460G MX1824G MX2472F MX74UP MX74UP MX1860F MX1842F MX2472F MX1836G MX54P MX2460F MX2424F MX2448F MX2436F MX1848F MX2436G MX2472G MX13P MX33P MX2430F MX2436F MX2442F MX1854F MX63P MX2430F MX1872F MX54UP MX86UP MX1836G
82003-204EA 291.03 USD InStock
82003-204 97005-482 19766-280 82003-246 97005-484 82003-208 82003-206 82003-252 19766-288 89369-948 82003-250 89369-946 82003-256 89369-944 82003-254 82003-238 19766-270 97005-864 97005-866 82003-244 82003-242 82003-226 CA89369-940 89369-930 82003-224 82003-228 97005-862 CA89369-936 82003-230 CA89369-934 CA89369-932 82003-234 19766-262 CA89369-930 82003-232 19766-228 CA89369-938 89369-942 82003-216 89369-940 82003-258 82003-214 82003-218 CA89369-948 89369-938 CA89369-946 89369-936 CA89369-944 19766-330 82003-222 89369-934 82003-220 CA89369-942 89369-932
MetroMax i™ Shelves and Posts, Metro™
Furniture Shelves
Microban® antimicrobial product protection built in to the shelf mats to inhibit stain and odor causing bacterial growth.
  • Posts feature polymer construction and are adjustable at 2.5 cm (1") increments
  • Solid mats are excellent for spill containment
Shelves consist of a shelf frame, open-grid or solid shelf mat, and four wedge connectors. Open-grid mats allow for air circulation and reduce dust and moisture buildup. Shelf mats may be removed from the shelf frame for easy cleaning.
Shelves may be adjusted in 2.5 cm (1") increments on the shelf posts (sold separately). Shelves up to 121.9 cm (48") in length support up to 362.9 kg (800lbs.) per level. Shelves 137.2 (54") and longer can support up to 272.2 kg (600 lbs.) per level. MetroMax i™ products are ideal for high-moisture applications and are recommended for continuous use in temperatures from –29 to +52 °C (–20 to +125 °F), with acceptable intermittent exposure to 93 °C (200 °F).
Stationary posts are fitted with adjustable leveling bolts to compensate for uneven surfaces. Mobile posts are fitted to accept casters (sold separately).
Ordering information: MetroMax i™ products feature a lifetime manufacturer's warranty against corrosion and rust. For additional accessories such as shelf dividers, stackable ledges, tray slides, label holders, and more, see 82027-268 series. For casters, see 19766-254 series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty