Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.0%, VWR® ACS, Reagent Grade for histology

Supplier: VWR International
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-Propanolsec-Propyl Alcohol2-Hydroxy propaneDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

ES34119VWR
89370-086CS 213.53 USD InStock
89370-086
2-Propanol ≥99.0%, VWR® ACS, Reagent Grade for histology
2-Propanol
Ideal for specialty fixation and dehydration in the histology laboratory.

  • Can be used as a substitute for ethyl alcohol

Isopropyl alcohol is recommended for small tissue and biopsy processing.


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
USP Assay (by GC) (corrected for water) min. 99.0%
ACS Assay (by GC) (corrected for water) min. 99.5%
USP Identification A To Pass
USP Identification B To Pass
ACS Color (APHA) max.10
ACS Appearance Clear Liquid
ACS Solubility in Water Solution as clear as equal volume of water
ACS Residue After Evaporation max.
USP Limit of Non-Volatile Residue max. 2.5 mg/50 ml
USP Limit of Volatile Impurities
Diethyl Ether max. 0.1%
Acetone max. 0.1%
Diisopropyl Ether max. 0.1%
n-Propyl Alcohol max. 0.1%
2-Butanol max. 0.1%
Total max. 1.0%
ACS Titrable Acid or Base max. 0.0001 meq/g
USP Acidity NMT 0.70 mL of 0.02N NaOH required for neutralization
ACS Water max. 0.2%
USP Water Determination max. 0.5%
ACS Carbonyl Compounds 0.002% max. each of propionaldehyde or acetone max. 0.002%
USP Refractive Index @ 20°C 1.376-1.378
USP Specific Gravity @25°C 0.783-0.787

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty