Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
GLASS TUBING CUTTER GRIFFIN
Supplier: HUMBOLDT MFG CO
Glass Tubing Cutter, Griffin-type^ Constructed of nickel-plated zinc alloy die-casting. Cuts tubing up to 38mm in diameter. Spring-loaded to stay open until used. Replaceable hardened steel cutting wheel.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLASS TUBING CUTTER GRIFFIN 89370-260 H-13520 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty