Would you like to log in again?
CLAMP BUNSEN ROUND JAW EX LRG
Supplier: HUMBOLDT MFG CO
Bunsen Round Jaw Extension Clamp^ Clamps are made of heavy-gauge stamped steel and have plastisol-coated jaws. Jaw opening: 18 to 51mm. Length: 229mm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLAMP BUNSEN ROUND JAW EX LRG 89370-262 H-8572 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty