Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Support Rings, Wire, Humboldt
Supplier: Humboldt
H-18325 H-18355 H-18335 H-18345
89370-318EA 64.29 USD InStock
89370-318 89370-314 89370-316 89370-312
Support Rings, Wire, Humboldt
Support Rings
Formed, bright basic wire circle is welded to shank (except 2" size)
Aluminum muff. Attaches to support rods up to 3/4" (19 mm) in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty