Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Support Rings, Cast Iron, Humboldt Manufacturing
Supplier: Humboldt
H-18370 H-18331 H-18350 H-18340 H-18320 H-18360
89370-304EA 54.34 USD InStock
89370-304 89370-306 89370-322 89370-310 60120-088 60120-022
Support Rings, Cast Iron, Humboldt Manufacturing
Support Rings
Distance from the center of the ring to the center of the clamp is 108mm ( 41/4")
Rings are plated and attach to 13mm (1/2") diameter rods.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty