Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish) alcoholic solution, Shandon™ cytoplasmic counterstain

Supplier: Epredia
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

6766008 6766007 6766008
89370-828CS 196.02 USD InStock
89370-828 89212-376 CA89212-376
Eosin Y (yellowish) alcoholic solution, Shandon™ cytoplasmic counterstain
Eosin Y (yellowish)
The Epredia™ Shandon™ Eosin-Y Staining Solutions are fast-acting cytoplasmic counterstain.

  • Stable and precipitate-free; no filtering necessary
  • Produces consistent results
  • pH-adjusted for maximum stain uptake
  • Produces brilliant staining
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic and aqueous formulations
  • Ready-to-use


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty