Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Four Student Cabinet Tables Construced of Solid Oak with Locking Doors and Drawers

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
2404K 2402K C2401K 2406K C2406K 2401K
89374-896EA 6272.59 USD InStock
89374-896 89374-890 89374-912 89374-904 89374-914 89374-888
VWR® Four Student Cabinet Tables Construced of Solid Oak with Locking Doors and Drawers
Furniture Tables and Workbenches
Four student cabinet table constructed of solid oak, oak and hardwood veneers features a center cabinet with adjustable shelves, locking doors and four locking drawers.

  • ¾" phenolic resin surface
  • Finishing the desk is rubber base molding, leg boots and a durable chemical resistant UV finish
  • This cabinet table allows workspace and storage for four users

This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. These tables are designed for use at sitting height. This unit is available with our complete line of top materials to satisfy every requirement or budget.

Certifications: It is SEFA compliant and MAS certified green.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty