Would you like to log in again?
VWR® Four Student Perimeter Workstation Made from Solid Oak
Supplier: VWR
VWR®
C2306K
89374-910EA 7513.33 USD InStock
89374-910
VWR® Four Student Perimeter Workstation Made from Solid Oak
Furniture Tables and Workbenches
Four student cabinet table constructed of solid oak, oak and hardwood veneers features a center cabinet with adjustable shelves, book compartments in the aprons, and locking doors.
  • 1" solid epoxy resin surface
  • Four student workstation features a locking center cabinet with adjustable shelf
  • Includes four book compartments in the aprons
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. The center cabinet is available with two locking drawers or as a plain base cabinet with adjustable shelf. These tables are designed for use at sitting height.
Certifications: It is SEFA compliant and MAS certified green.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty