Would you like to log in again?
VWR® Forward Vision III Workstation Flat Top for Two Students
Supplier: VWR
VWR®
2926K
89374-920EA 11742.03 USD InStock
89374-920
VWR® Forward Vision III Workstation Flat Top for Two Students
Furniture Tables and Workbenches
Two student workstation constructed of solid oak, oak and hardwood veneers.
  • 1" Solid epoxy resin surface
  • Constructed of solid oak and oak veneers
  • Designed to be used by two students
  • Finishing the workstation is rubber base molding and a durable chemical resistant UV finish
  • Full cabinet doors with locks include adjustable shelves
  • Includes a sink and two combination cold water/gas fixtures
  • One GFI protected AC duplex receptacle
Durable lab table features a large 68×36" tabletop. It comes standard with two combination cold water/gas fixtures and one GFI AC electrical receptacle. Your choice of two cabinets: Full door cabinet with adjustable shelves or door/drawer cabinet with a drawer and storage area with one adjustable shelf. They are finished with a chemical resistant, earth-friendly UV finish. An 18" wide removable panel is located between the storage cabinets and provides access to all utilities. Unit can be ordered with a drop-in sink or with a flat top without sink or fixtures. Assembly and installation of electricity and plumbing is required.
Sink dimension: 12W×8L×6"D
Certifications: SEFA compliant and MAS certified green.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty