Would you like to log in again?
VWR® Two Student Book Compartment Table/Workstation with Cabinet
Supplier: VWR
VWR®
C2716KF
89374-990EA 5633.94 USD InStock
89374-990
VWR® Two Student Book Compartment Table/Workstation with Cabinet
Furniture Tables and Workbenches
Two student single faced workstation that provides a fully equipped science center.
  • 1" solid epoxy resin surface
  • Includes two book compartments in the aprons
  • The cabinets have locking cupboard
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
Single faced tables that provide a fully equipped science center from two to eight students. Each unit is constructed of solid oak, and hardwood veneers. The single faced units contain a solid epoxy resin sink along with cold water faucets for every two students. All units contain locks for the drawers and doors. Choose from models with a full cupboard only or a half cupboard and dovetailed drawers.
Sink dimension: 12W×8L×6"D
Ordering information: Can be ordered without sink, fixtures and rod sockets. Two student single faced workstation includes a sink, two cold water/gas fixture, a GFI protected AC duplex receptacle and two rod sockets.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty