Would you like to log in again?
VWR® Perimeter Four Student Single Faced Workstation With Science Center
Supplier: VWR
VWR®
C2726K
89374-992EA 18098.11 USD InStock
89374-992
VWR® Perimeter Four Student Single Faced Workstation With Science Center
Furniture Tables and Workbenches
Four student single faced workstation that provides a fully equipped science center.
  • 1" solid epoxy resin surface
  • Includes four book compartments in the aprons
  • The cabinets have locking cupboard
  • The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
The single faced units contain a solid epoxy resin sink along with cold water faucets for every two students. Every unit contains enough gas cocks for each student. All units contain locks for the drawers and doors. Choose from models with a full cupboard only or a half cupboard and dovetailed drawers. Add 66" in length for each additional two students. Your choice of 1" epoxy resin or ¾" phenolic resin tops.
Sink dimension: 12W×8L×6"D
Certifications: It is SEFA compliant and MAS certified green.
Ordering information: Four student single faced workstation includes two sinks, four cold water/gas fixtures, two GFI protected AC duplex receptacles and four rod sockets. Can be ordered without sink, fixtures and rod sockets.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty