Would you like to log in again?
HyClone™ HyCryo-Stem Cryopreservation Media, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
SR30002.02 SR30002.02
89399-934EA 151.66 USD InStock
CA89399-934 89399-934
HyClone™ HyCryo-Stem Cryopreservation Media, HyClone products (Cytiva)
Cell Freezing Media
HyClone™ HyCryo-STEM is a serum-free cryopreservation media designed specifically to maintain differentiation potential and minimise spontaneous differentiation of stem cells.
  • Greater recovery of viable neural progenitor stem cells compared to industry standard cryopreservation media (growth media containing 10% DMSO)
  • Achieves similar recovery of viable human stem cells compared to the industry standard cryopreservation media (a cocktail of 25% serum replacement and 10% DMSO)
Note: For further manufacturing or research use. Not for diagnostic or therapeutic use.
Delivery information: Supplied as a 2X solution, 0,2 µm sterile filtered.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty