Would you like to log in again?
STRETCHSTOP - ANSI Z7
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
STRETCHSTOP - ANSI Z7^ Miller StretchStop* single leg lanyard with SofStop* shock absorber and ANSI Z-359 snap hooks. 6 ft in length.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STRETCHSTOP - ANSI Z7 89400-460 913SS-Z7/6FTGN Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty