Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Miller® Turbo T-BAK Personal Fall Limiters, Honeywell Safety
Supplier: Miller by Honeywell
Miller®
MFLT-4/7.5FT MFLT-1/7.5FT MFLT-5/7.5FT MFLT-3/7.5FT MFLT-2/7.5FT
89400-470EA 382.03 USD InStock
89400-470 89400-472 89400-474 89400-466 89400-468
Miller® Turbo T-BAK Personal Fall Limiters, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
The Miller® Turbo T-BAK was designed to tie-back anywhere along the lifeline and is therefore the only product on the market that meets the ANSI Z359.14. Standard and provides a continuous 100% tie-off protection.
  • Cost-effective alternative to tie-back, shock-absorbing lanyards
  • All-in-one design reduces equipment required on the job site
  • Anchors below a user's harness back D-ring for greater safety
  • Integrated shock absorber especially designed for tie-back applications with load indicator device
  • Working capacity: 7.5' (2.3 m)
  • Heavy-duty web lifeline smoothly extends/retracts without interruption
Engineered with high-tech strength and abrasion-resistant webbing, the turbo T-BAK provides more tie-off anchorage locations with a greater variety of beam-size options. It also simplifies user training for improved safety.
Certifications: Meets all applicable OSHA, ANSI, CSA and CE standards, including the ANSI Z359.14-2012 requirement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty