Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Miller® Falcon™ Self-Retracting Lifeline, Honeywell Safety
Supplier: Miller by Honeywell
Miller® Falcon
MP30G-Z7/30FT MP65G-Z7/65FT MP16P-Z7/16FT MP20P-Z7/20FT MP20G-Z7/20FT MP50G-Z7/50FT
89400-528EA 1192.93 USD InStock
89400-528 89400-530 89400-522 89400-532 89400-524 89400-526
Miller® Falcon™ Self-Retracting Lifeline, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
Nearly indestructible, the Miller® Falcon™ Self-Retracting Lifeline is built with a high impact-resistant nylon housing that withstands the harsh rigors of the toughest applications. Its smooth, round edges increase durability.
  • Unique side payout design reduces wear on the entry guide bushing and cable for smooth operation with less drag
  • Smooth, quiet operation; Engineered spring retraction tension is designed not to pull or drag on a worker to improve productivity and reduce fatigue
  • Built-in, ergonomically-designed molded handle on the 50' (15 m) and 65' (20 m) cable units provide support and leverage when attaching to anchorage connector
  • Dual swivels (unit top and snap hook) included on all cable models to minimize lifeline from twisting during use
  • New and improved load indicators identify when the unit needs to be removed from service
  • Cable models rated to 310 lbs. (140.6 kg) capacity, or 400 lbs. (181.4 kg) when using a SofStop shock absorber (928LS/18INBK)
  • Web models rated for up to 400 lbs. (181.4 kg) worker
Load indicators on web models feature a flag on the webbing. Load indicators on cable models are on the snap hook. Units are repairable and require no annual factory recertification, which lowers the cost of ownership and increases productivity (except in Canada where the Canadian Standards Association requires annual factory recertification).
Certifications: Meets all applicable OSHA 1926.502, ANSI A10.32, ANSI Z359.14 Class B, and CSA Z259.2.2 requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty