Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Miller® MightyLite™ Self Retracting Lifeline, Honeywell Safety

Supplier: Miller by Honeywell
Miller®
RL50G-Z7/50FT RLS30G-Z7/30FT RL20P-Z7/20FT RL65G-Z7/65FT RL20G-Z7/20FT RL50P-Z7/50FT
89400-538EA 1129.12 USD InStock
89400-538 89400-540 89400-542 89400-544 89400-534 89400-536
Miller® MightyLite™ Self Retracting Lifeline, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
Miller® MightyLite™ Self-Retracting Lifelines (SRL’s) are up to 30% lighter than competitive models in their class. Corrosion-resistant stainless steel and aluminum housing extend service life.

  • Durable and dependable, corrosion-resistant stainless steel and aluminum housing extend service life
  • No annual factory recertification required
  • Rated for up to a 310 lb. (140.6 kg) user
  • Available in 20' (6 m), 30' (9 m), 50' (15 m), 65' (20 m), 100' (30 m), 130' (40 m) and 175' (53 m) lengths

Certifications: Meets OSHA 1926.502, 1910.66, ANSI A10.32, *ANSI Z359.14-2012 Class B and CSA Z259.2.2-98 Type 2 requirements.

Delivery information: Carabiner and tagline included*

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty