Would you like to log in again?
SKYGRIP 60FT HLL I-BEAM EA1
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
SKYGRIP 60FT HLL I-BEAM EA1^ Miller Skygrip System for steel application with standard base. Fit beam fl anges 4-in. to 18-in. (.1 m to .5 m) wide and up to 2-1/4-in. (57 mm) thick. 60-ft. (18 m) system
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SKYGRIP 60FT HLL I-BEAM EA1 89400-584 SGS18/60FT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty