Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Miller® Durahoist™ Portable Confined Space Systems, Honeywell Safety
Supplier: Honeywell Safety
Miller®
DH-1/ DH-2/
89400-612EA 12038.46 USD InStock
89400-612 89400-614
Miller® Durahoist™ Portable Confined Space Systems, Honeywell Safety
Fall Protection Fall Arresters
The Miller DuraHoist™ portable confined space system is designed for manhole and confined space entry/retrieval, and fall arrest applications.
  • System breaks down into four lightweight aluminum components
  • No tools required for assembly
The modular design allows the use of components in various configurations and applications, such as fixed and portable bases and an optional mast with extensions. Multiple winches and/or lifeline systems may be mounted as required. The lightweight, manageable components are easy to store, transport, and set-up.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty