Would you like to log in again?
GLIDE-LOC KIT GALV 30FT EA1
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
GLIDE-LOC KIT GALV 30FT EA1^ Galvanized GlideLoc kit 30ft. Kits come complete with GlideLoc rail, gated top end-stop, gated bottom end-stop, ladder attachment brackets, and hardware. Systems accommodate one worker per section of rail. GlideLoc fall arresters are sold separately.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLIDE-LOC KIT GALV 30FT EA1 89400-634 GG0030 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty