Would you like to log in again?
EYEWEAR WRAP-ARND GRAY FRM/BLUE MIR LENS
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Safety glasses grey frame^ Safety glasses, Soft nosepiece, delivers ultra-light-weight, sport inspired protection, 9-base polycarbonate hardcoated lenses provide a 180 deg field of distortion-free vision, Frame colour: Grey, Lens colour: Blue mirror, Temple Length: 115mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EYEWEAR WRAP-ARND GRAY FRM/BLUE MIR LENS 89400-865 A803 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty