Would you like to log in again?
Calibration Baths, PolyScience
Supplier: Polyscience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AP15RCAL-A11B PD15HCAL-A12E PP15HCAL-A11B PD15HCAL-A11B PP15HCAL-A12E
89400-936EA 6766.42 USD InStock
89400-936 89400-938 89400-940 89400-942 89400-944
Calibration Baths, PolyScience
Circulating Thermostats Open Circulator Systems Calibration Circulators
PolyScience Calibration Baths accommodate up to 6 temperature measuring devices and come with 2-point factory calibration, as well as bath lids with sleeves for two 3mm, 4mm and 6mm diameter openings
Calibration Baths are available in Heated and Refrigerated/Heated models. Swivel 180™, LidDock™ and DuraTop™ features all come standard. WhisperCool® Environmental Control System is standard on refrigerated units. All models include USB-A, RS-232, RS-485 and Ethernet, and include an external temperature probe port.
Event scheduling (time and date), along with real-time clock and temperature trends (for up to 10 days) included. Drain ports make it easy to remove liquids from the bath and the washable air filter helps keep the circulator energy-efficient and running at peak performance.
Available with the Performance Programmable (OpenMode programming for unlimited steps and programs, for specific test protocols) or Performance Digital Controller, all Calibration Baths feature an icon-driven SmartTouch interface, which leads you through menus, functions and options, enabling you to make any adjustments, even with your gloves on. All screens and menus, including on-screen help, can be displayed in any of 11 different languages.
Certifications: Complies with DIN 12876-1 Class III safety requirements for use with flammable liquids, cETLus compliant.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty