Would you like to log in again?
Zirconium Crucibles, Kitco Metals Inc.
Supplier: XRF Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ZXS25 ZXS75 ZXS30 ZXS35 ZXS100 ZXS45 ZXS50
89401-348EA 140.7 USD InStock
89401-348 89401-346 89401-356 89401-344 89401-354 89401-352 89401-360
Zirconium Crucibles, Kitco Metals Inc.
Crucibles
Zirconium's distinctive properties make it a popular choice for fusions and fluxes in analytical chemistry and chemical processing.
These standard shape Zirconium crucibles are highly resistant to corrosion. They do not require handling with platinum tipped tongs and they can be cleaned with most common agents (except hydrofluoric acid). Zirconium crucibles suffer little to no attack or oxidation and are resistant to melts of alkali carbonates, hydroxides, peroxides, borates, nitrate chlorides, and some fluorides. This construction virtually eliminates the problem of sample contamination. Lids are available upon request.
Ordering information: Lids are sold separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty