Would you like to log in again?
Low Form Zirconium Crucibles, Kitco Metals Inc.
Supplier: XRF Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ZLF25 ZLF45 ZLF30 ZLF50 ZLF100 ZLF20 ZLF35
89401-368EA 120.8 USD InStock
89401-368 89401-378 89401-386 89401-374 89401-372 89401-370 89401-380
Low Form Zirconium Crucibles, Kitco Metals Inc.
Crucibles
Zirconium's distinctive properties make it a popular choice for fusions and fluxes in analytical chemistry and chemical processing.
These low form Zirconium crucibles are highly resistant to corrosion, and do not require handling with platinum tipped tongs. They can be cleaned with most common agents (except hydrofluoric acid), and suffer little to no attack or oxidation. Zirconium crucibles are resistant to melts of alkali carbonates, hydroxides, peroxides, borates, nitrate chlorides, and some fluorides. This construction virtually eliminates the problem of sample contamination. Lids are available upon request.
Ordering information: Lids are sold separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty