Would you like to log in again?
COLUMN FLASH SS 120G PK5 SCR
Supplier: Agela Technologies
Screw-on Flash Column spacer luer lock^ For inlet and outlet Spherical Standard Silica(CS) I-series 20-45 um 60 120g 5/Pack
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN FLASH SS 120G PK5 SCR 89401-834 SN-SS130120-0 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty